Learn More
Apexbio Technology LLC ALLYL-METHYL-SULFIDE-200MG
SDP
Catalog No. 5000569369
Encompass_Preferred
Allyl methyl sulfide (CAS No 10152-76-8) is an organosulfur compound primarily studied in biomedical research for its bioactive properties Originating from garlic it is known for its potential antioxidant and antimicrobial activities The compound acts by modulating oxidative stress pathways and inhibiting microbial growth potentially impacting cellular signaling processes In vitro it exhibits activity in the nanomolar to low micromolar range showcasing its potency across various experimental conditions Common applications include studies involving cell lines and animal models to explore its therapeutic potential and utility in drug screening assays Experimental concentrations typically depend on the specific research design allowing for flexibility in diverse investigative contexts
Catalog No. 50-005-69369 Supplier Apexbio Technology LLC Supplier No. C7172200
Allyl methyl sulfide (CAS No 10152-76-8) is an organosulfur compound primarily studied in biomedical research for its bioactive properties Originating from garlic it is known for its potential antioxidant and antimicrobial activities The compound acts by modulating oxidative stress pathways and inhibiting microbial growth potentially impacting cellular signaling processes In vitro it exhibits activity in the nanomolar to low micromolar range showcasing its potency across various experimental conditions Common applications include studies involving cell lines and animal models to explore its therapeutic potential and utility in drug screening assays Experimental concentrations typically depend on the specific research design allowing for flexibility in diverse investigative contexts