Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Allyl-PEG2-OH | 15075-50-0 | 99.3% | 146.18 g/mol | C7H14O3 | 250 MG
SDP

Catalog No. 5000209274
Encompass_Preferred

Allyl-PEG2-OH is an allyl-terminated polyethylene glycol (PEG) derivative with a terminal hydroxyl group used as a short PEG linker in chemical synthesis, bioconjugation, and drug-delivery research. It is a low molecular weight PEG derivative commonly handled under cold storage conditions to preserve stability.

  • Allyl-terminated polyethylene glycol linker with a terminal hydroxyl group.
  • Molecular formula C7H14O3 and molecular weight 146.18 g/mol.
  • High purity (GC) 99.33%.
  • Suitable for chemical synthesis, bioconjugation, and drug-delivery linker design.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-09274 Supplier Medchemexpress LLC Supplier No. 250MGHYW047691
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allyl-PEG2-OH is an allyl-terminated polyethylene glycol (PEG) derivative with a terminal hydroxyl group used as a short PEG linker in chemical synthesis, bioconjugation, and drug-delivery research. It is a low molecular weight PEG derivative commonly handled under cold storage conditions to preserve stability.

  • Allyl-terminated polyethylene glycol linker with a terminal hydroxyl group.
  • Molecular formula C7H14O3 and molecular weight 146.18 g/mol.
  • High purity (GC) 99.33%.
  • Suitable for chemical synthesis, bioconjugation, and drug-delivery linker design.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.