Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Phenoxyacetate 98.0+%, TCI America™
SDP

Catalog No. A217725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A217725G 25 g
A2177250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A217725G Supplier TCI America Supplier No. A217725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7493-74-5
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00044632
InChI Key VUFZVGQUAVDKMC-UHFFFAOYSA-N
Synonym allyl phenoxyacetate, allyl 2-phenoxyacetate, acetic acid, phenoxy-, 2-propenyl ester, acetate p.a., acetate pa, phenoxyacetic acid allyl ester, 2-propenyl phenoxyacetate, unii-q3p8uaf9we, acetic acid, phenoxy-, allyl ester, acetate p.a
PubChem CID 24117
IUPAC Name prop-2-enyl 2-phenoxyacetate
SMILES C=CCOC(=O)COC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 137°C
Quantity 25 g
UN Number 2810
Formula Weight 192.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Phenoxyacetate
EINECSNumber (3)-0633
RTECSNumber AJ2240000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.