Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allyl phenyl sulfone

Catalog No. 0606150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-061-50ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-061-50ML Supplier Sigma Aldrich Supplier No. 0606150ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,N,N-Trimethylaniline is a tertiary amine that can undergo iron-catalyzed oxidative C-C coupling with phenylacetylene and benzamide in the presence of tert-butylperoxide to form N,4-dimethyl-N-(3-phenylprop-2-ynyl)benzenamine and N-((methyl(p-tolyl)amino)methyl)benzamide, respectively.

Specifications

CAS 16212-05-8
Density 1.189 g/mL (at 25°C (literature))
Boiling Point 110°C to 113°C (0.5 mmHg)
Molecular Formula C9H10O2S
Refractive Index n20/D 1.548 (literature); n20/D 1.548
Linear Formula C6H5SO2CH2CH=CH2
MDL Number MFCD00014739
Quantity 500 mL
Molecular Weight (g/mol) 182.24
Percent Purity ≥98.0% (GC)
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.