Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl(tert-butyl)dimethylsilane 97.0+%, TCI America™
SDP

Catalog No. A22985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22985G Supplier TCI America Supplier No. A22985G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 74472-22-3
Molecular Formula C9H20Si
Molecular Weight (g/mol) 156.344
InChI Key KALUNQGXJIFEDG-UHFFFAOYSA-N
PubChem CID 10511119
IUPAC Name tert-butyl-dimethyl-prop-2-enylsilane
SMILES CC(C)(C)[Si](C)(C)CC=C

Specifications

Color Colorless
Boiling Point 152°C
Quantity 5 g
UN Number 1993
Formula Weight 156.34
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl(tert-butyl)dimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.