Learn More
Enzo Life Sciences Allylamine dUTP (2.5µmol)
SDP
Catalog No. 502012558
Encompass_Preferred
Allylamine-2’-deoxyuridine-5’-triphosphate can replace TTP in reactions where it serves as a substrate for E. coli DNA polymerase 1 (holoenzyme and Klenow fragment), terminal deoxynucloetide transferase, T4 and Taq DNA polymerases, and reverse transcriptases (from AMV and M-MuLV). Allylamine-dUTP is incorporated into DNA by a variety of labeling reactions including nick translation, random primed DNA synthesis and 3’ end labeling reactions. Allylamine-labeled DNA can be efficiently conjugated to the active ester form of signal generating moieties such as fluorescent dyes, biotin and other haptens to produce labeled probes for hybridization/detection assays. The efficient incorporation of Allylamine-dUTP, in contrast to many dye-labeled nucleotides, provides for greater incorporation of signaling moieties and stronger signals. Alternative name: Allylamino-2'-deoxyuridine-5'-triphosphate. Concentration: 50mM. Purity: ≥93% (HPLC). Formulation: Liquid. Solution in water.
Catalog No. 50-201-2558 Supplier Enzo Life Sciences Supplier No. ENZ42861
Allylamine-2’-deoxyuridine-5’-triphosphate can replace TTP in reactions where it serves as a substrate for E. coli DNA polymerase 1 (holoenzyme and Klenow fragment), terminal deoxynucloetide transferase, T4 and Taq DNA polymerases, and reverse transcriptases (from AMV and M-MuLV). Allylamine-dUTP is incorporated into DNA by a variety of labeling reactions including nick translation, random primed DNA synthesis and 3’ end labeling reactions. Allylamine-labeled DNA can be efficiently conjugated to the active ester form of signal generating moieties such as fluorescent dyes, biotin and other haptens to produce labeled probes for hybridization/detection assays. The efficient incorporation of Allylamine-dUTP, in contrast to many dye-labeled nucleotides, provides for greater incorporation of signaling moieties and stronger signals. Alternative name: Allylamino-2'-deoxyuridine-5'-triphosphate. Concentration: 50mM. Purity: ≥93% (HPLC). Formulation: Liquid. Solution in water.