Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allylboronic Acid Pinacol Ester

Catalog No. 3246471G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-464-71G 1 g
32-464-710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 32-464-71G Supplier Sigma Aldrich Supplier No. 3246471G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used forPalladium-catalyzed Suzuki-Miyaura cross-coupling reactions and olefin metathesis Intermolecular radical additions Allylboration of aldehydes catalyzed by chiral spirobiindane diol (SPINOL) based phosphoric acids Cobalt-catalyzed regioselective hydrovinylation of dienes with alkenes Nucleic acid-templated energy transfer leading to a photorelease reaction Stereoselective indium-catalyzed Hosomi-Sakurai reactions Reagent used in Preparation ofCyclic sulfone hydroxyethylamines as BACE1 inhibitors for reduction of Amyloid +a-Peptides Transmetalation of carbene Ru iodide with Ag carboxylates to give C-H-activated Ru carbene complexes as catalysts for Z-selective olefin metathesis Allylboronation reagent for the preparation of allylic alcohols, and homoallylic amines.

Specifications

CAS 72824-04-5
Density 0.896 g/mL at 25°C
Boiling Point 50°C to 53°C (5 mmHg, lit.)
Molecular Formula C9H17BO2
Refractive Index n20/D 1.4268
Linear Formula ((CH3)4C2O2)BCH2CH=CH2
MDL Number MFCD00013347
Quantity 1 g
Synonym 2-Allyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; 4,4,5,5-tetramethyl-2-(2-propen-1-yl)-1,3,2-dioxaborolane; 4,4,5,5-tetramethyl-2-(2-propenyl)-1,3,2-dioxaborolane; Allyl pinacol boronate; Pinacol allylboronate; Pinacolyl 2-propenylboronate
Molecular Weight (g/mol) 168.04 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.