Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylcyclopentane, 97%

Catalog No. AAB2022806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2022806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2022806 Supplier Thermo Scientific Chemicals Supplier No. B2022806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3524-75-2 | C8H14 | 110.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3524-75-2
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
MDL Number MFCD00001390
InChI Key NHIDGVQVYHCGEK-UHFFFAOYSA-N
Synonym allylcyclopentane, 3-cyclopentylpropene, cyclopentane, 2-propenyl, cyclopentane, allyl, 3-cyclopentyl-1-propene, allyl cyclopentane, 2-propenylcyclopentane, 1-propene, 3-cyclopentyl, 3-cyclopentylprop-1-ene
PubChem CID 77059
IUPAC Name prop-2-enylcyclopentane
SMILES C=CCC1CCCC1

Specifications

Melting Point -111°C
Density 0.793
Boiling Point 125°C to 127°C
Flash Point 12°C (53°F)
Refractive Index 1.441
Quantity 5 g
UN Number UN3295
Beilstein 2036419
Formula Weight 110.2
Percent Purity 98%
Chemical Name or Material Allylcyclopentane
Show More Show Less
Hazard Category H225-H302+H332
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 222-542-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.