Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyldimethylamine, MP Biomedicals

Catalog No. ICN214464 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN214464 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN214464 Supplier MP Biomedicals, Inc Supplier No. 0521446410

Chemical Identifiers

CAS 2155-94-4
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00038234
InChI Key GBCKRQRXNXQQPW-UHFFFAOYSA-N
Synonym n,n-dimethylallylamine, allyldimethylamine, dimethylallylamine, 2-propen-1-amine, n,n-dimethyl, 1-dimethylamino-2-propene, n-allyl-n,n-dimethylamine, n-allyldimethylamine, dimethyl prop-2-en-1-yl amine, allylamine, n,n-dimethyl, ch2=chch2n ch3 2
PubChem CID 75082
IUPAC Name dimethyl(prop-2-en-1-yl)amine
SMILES CN(C)CC=C

Specifications

Quantity 10 g
Formula Weight 85
Chemical Name or Material Allyldimethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.