Learn More
Strem, An Ascensus Company CAS# 12012-95-2. 10g. Allylpalladium chloride dimer, min. 98%. MFCD00044874
SDP
Catalog No. 502016794
Encompass
CAS# 12012-95-2. 10g. Allylpalladium chloride dimer, min. 98%. MFCD00044874. Molecular Weight: 365.86. Molecular Formula: [C3H5PdCl]2. Color/form: yellow xtl. Strem# 46-0100. http://www.strem.com/catalog/v/46-0100/
Catalog No. 50-201-6794 Supplier Strem, An Ascensus Company Supplier No. 46010010G
CAS# 12012-95-2. 10g. Allylpalladium chloride dimer, min. 98%. MFCD00044874. Molecular Weight: 365.86. Molecular Formula: [C3H5PdCl]2. Color/form: yellow xtl. Strem# 46-0100. http://www.strem.com/catalog/v/46-0100/