Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylpentafluorobenzene, 97%, Thermo Scientific™

Catalog No. AC310850050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC310850050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC310850050 Supplier Thermo Scientific Chemicals Supplier No. 310850050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1736-60-3
Molecular Formula C9H5F5
Molecular Weight (g/mol) 208.13
MDL Number MFCD00000301
InChI Key YBDBTBVNQQBHGJ-UHFFFAOYSA-N
Synonym allylpentafluorobenzene, benzene, allylpentafluoro, 3-pentafluorophenyl-1-propene, 2,3,4,5,6-pentafluoro allylbenzene, benzene, pentafluoro-2-propenyl, 1,2,3,4,5-pentafluoro-6-prop-2-en-1-yl benzene, allyl pentafluorobenzene, pubchem13366, acmc-1bth5
PubChem CID 74435
IUPAC Name 1,2,3,4,5-pentafluoro-6-prop-2-enylbenzene
SMILES FC1=C(F)C(F)=C(CC=C)C(F)=C1F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -64.0°C
Color Colorless
Density 1.3580g/mL
Boiling Point 148.0°C to 149.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4270 to 1.4290 (20°C, 589nm)
Linear Formula C6F5CH2CH=CH2
Quantity 5 g
Specific Gravity 1.358
Formula Weight 208.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Allylpentafluorobenzene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
EINECSNumber 217-083-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.