Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylsuccinic Anhydride 97.0+%, TCI America™
SDP

Catalog No. A12835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12835G 5 g
A128325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12835G Supplier TCI America Supplier No. A12835G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7539-12-0
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00085227
InChI Key WUMMIJWEUDHZCL-UHFFFAOYSA-N
Synonym 4-Pentene-1,2-dicarboxylic Anhydride, (2-Propenyl)butanedioic Anhydride
PubChem CID 111009
IUPAC Name 3-prop-2-enyloxolane-2,5-dione
SMILES C=CCC1CC(=O)OC1=O

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 2810
Boiling Point 143°C
Formula Weight 140.14
Percent Purity ≥97.0% (GC)
Chemical Name or Material Allylsuccinic Anhydride
EINECSNumber (5)-7013
RTECSNumber WN1050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.