Learn More
Medchemexpress LLC Allylthiourea | 109-57-9 | 100.0% | 500 G
SDP
Catalog No. 5000408884
Encompass_Preferred
Allylthiourea selectively inhibits the oxidation of ammonia and is commonly used to inhibit nitrification by targeting ammonia monooxygenase and chelating copper in the active site to suppress its activity. It also exhibits anticancer activity, showing cytotoxicity against the MCF-7 cell line with an IC50 of 5.22 mM.
  • Selectively inhibits ammonia oxidation
  • Inhibits nitrification by targeting ammonia monooxygenase
  • Chelates copper in the active site to suppress activity
  • Exhibits anticancer activity against MCF-7 cell line (IC50 of 5.22 mM)
  • Used in research for micropollutant biodegradability
  • Used in cancer studies
Catalog No. 50-004-08884 Supplier Medchemexpress LLC Supplier No. HYB0543500G
Allylthiourea selectively inhibits the oxidation of ammonia and is commonly used to inhibit nitrification by targeting ammonia monooxygenase and chelating copper in the active site to suppress its activity. It also exhibits anticancer activity, showing cytotoxicity against the MCF-7 cell line with an IC50 of 5.22 mM.
  • Selectively inhibits ammonia oxidation
  • Inhibits nitrification by targeting ammonia monooxygenase
  • Chelates copper in the active site to suppress activity
  • Exhibits anticancer activity against MCF-7 cell line (IC50 of 5.22 mM)
  • Used in research for micropollutant biodegradability
  • Used in cancer studies