Learn More
Medchemexpress LLC Allylthiourea | 109-57-9 | 99.95% | 25 G
SDP
Catalog No. 5000407078
Encompass_Preferred
Allylthiourea selectively inhibits ammonia oxidation by targeting ammonia monooxygenase and chelating copper. It also demonstrates anticancer activity against the MCF-7 cell line. This compound is useful for research in micropollutant biodegradability and cancer studies.
  • Selectively inhibits ammonia oxidizers
  • Decreases intracellular copper in Methylosinus trichosporium OB3b
  • Exhibits anticancer activity
  • Used in research related to micropollutant biodegradability and cancer
Catalog No. 50-004-07078 Supplier Medchemexpress LLC Supplier No. 25GHYB0543
Allylthiourea selectively inhibits ammonia oxidation by targeting ammonia monooxygenase and chelating copper. It also demonstrates anticancer activity against the MCF-7 cell line. This compound is useful for research in micropollutant biodegradability and cancer studies.
  • Selectively inhibits ammonia oxidizers
  • Decreases intracellular copper in Methylosinus trichosporium OB3b
  • Exhibits anticancer activity
  • Used in research related to micropollutant biodegradability and cancer