Learn More
Apexbio Technology LLC Allylthiourea 109-57-9 5g
SDP
Catalog No. 502240655
Encompass_Preferred
Allylthiourea (CAS 109-57-9) is a small molecule inhibitor recognized for its specificity in disrupting the metabolic pathway of ammonia oxidation It achieves this by selectively inhibiting the activity of ammonia-oxidizing microorganisms thereby blocking the conversion of ammonia to nitrite Owing to this targeted mechanism allylthiourea is extensively employed as a research tool in studies investigating nitrification processes microbial ecology and nitrogen cycling in various biological and environmental systems Its application facilitates the elucidation of ammonia oxidation dynamics and the role of ammonia-oxidizers in complex microbial communities
This item is not returnable. View return policy
Catalog No. 50-224-0655 Supplier Apexbio Technology LLC Supplier No. B18825000
Allylthiourea (CAS 109-57-9) is a small molecule inhibitor recognized for its specificity in disrupting the metabolic pathway of ammonia oxidation It achieves this by selectively inhibiting the activity of ammonia-oxidizing microorganisms thereby blocking the conversion of ammonia to nitrite Owing to this targeted mechanism allylthiourea is extensively employed as a research tool in studies investigating nitrification processes microbial ecology and nitrogen cycling in various biological and environmental systems Its application facilitates the elucidation of ammonia oxidation dynamics and the role of ammonia-oxidizers in complex microbial communities