Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Allylthiourea, 98%
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 70.0°C to 78.0°C |
| Color | White to Beige |
| Loss on Drying | 1.5% max. (50°C, 2 hrs) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Molecular Formula | C4H8N2S |
| Linear Formula | H2C=CHCH2NHCSNH2 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. |
| Precautionary Statement | GHS P Statement Wash face,hands and any exposed skin thoroughly after handling. Do not eat,drink or smoke when using this product. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective |
| EINECSNumber | 203-683-5 |
| RTECSNumber | YR8050000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction