Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylthiourea, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
50g
100g
500g
109-57-9
C4H8N2S
116.18
MFCD00004940
HTKFORQRBXIQHD-UHFFFAOYSA-N
allylthiourea,n-allylthiourea,thiosinamine,1-allyl-2-thiourea,rhodalline,thiosinamin,tiosinamine,1-allylthiourea,aminosin,rhodallin
1549517
CHEBI:74079
prop-2-enylthiourea
C=CCNC(=S)N
109-57-9
C4H8N2S
116.18
MFCD00004940
HTKFORQRBXIQHD-UHFFFAOYSA-N
allylthiourea,n-allylthiourea,thiosinamine,1-allyl-2-thiourea,rhodalline,thiosinamin,tiosinamine,1-allylthiourea,aminosin,rhodallin
1549517
CHEBI:74079
prop-2-enylthiourea
C=CCNC(=S)N
Melting Point 70.0°C to 78.0°C
Packaging Glass bottle
Quantity 500g
Infrared Spectrum Authentic
Color White to Beige
Loss on Drying 1.5% max. (50°C, 2 hrs)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2C=CHCH2NHCSNH2
Beilstein 04, 211
Merck Index 15, 9515
Solubility Information Solubility in water: 67g/L (20°C). Other solubilities: 1370g/L methanol (20°C)- insoluble in benzene,soluble in ether,solutions of borax,benzoates,and urethane,slightly soluble in 70% alcohol
Formula Weight 116.18
Physical Form Powder, Crystals, Granules or Chunks
Percent Purity 98%
Chemical Name or Material Allylthiourea, 98%
Show More Show Less

GHS H Statement
Toxic if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 203-683-5

RTECSNumber : YR8050000

TSCA : TSCA