Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylthiourea, 98%, Thermo Scientific™

Catalog No. AC148802500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC148802500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC148802500 Supplier Thermo Scientific Chemicals Supplier No. 148802500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-57-9
Molecular Formula C4H8N2S
Molecular Weight (g/mol) 116.182
InChI Key HTKFORQRBXIQHD-UHFFFAOYSA-N
Synonym allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
PubChem CID 1549517
ChEBI CHEBI:74079
IUPAC Name prop-2-enylthiourea
SMILES C=CCNC(=S)N

Specifications

Melting Point 70.0°C to 78.0°C
Linear Formula H2C=CHCH2NHCSNH2
Quantity 250 g
Solubility Information Solubility in water: 67g/L (20°C). Other solubilities: 1370g/L methanol (20°C) - insoluble in benzene, soluble in ether, solutions of borax, benzoates, and urethane, slightly soluble in 70% alcohol
Formula Weight 116.18
Chemical Name or Material Allylthiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.