Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allyltributylstannane

Catalog No. 271411100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-141-1100G 100 g
27-141-15G 5 g
27-141-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 27-141-1100G Supplier Sigma Aldrich Supplier No. 271411100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Allylation reagent for aldehydes catalyzed by chiral Lewis acids and ytterbium(III) trifluoromethanesulfonate (cat. no. 430595). Undergoes tetrakis(triphenylphosphine)palladium(0) (cat. no. 216666) catalyzed coupling with iodoquinones and allyl acetates.Efficient allylation of aldehydes leading to homoallylic alcohols with trichloro-1,3,5-triazene.

Specifications

CAS 24850-33-7
Density 1.068 g/mL at 25°C
Boiling Point 88°C to 92°C (0.2 mmHg, lit.)
Molecular Formula C15H32Sn
Refractive Index n20/D 1.486
Linear Formula CH2=CHCH2Sn(CH2CH2CH2CH3)3
MDL Number MFCD00010346
Quantity 100 g
Synonym Allyltri-n-butyltin; Allyltributyltin; Tributyl-2-propenylstannane
Molecular Weight (g/mol) 331.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.