Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allyltrimethoxysilane

Catalog No. 4469555ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-695-55ML 5 mL
44-695-525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 44-695-55ML Supplier Sigma Aldrich Supplier No. 4469555ML
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Allyltrimethoxysilane is an allylating reagent that can be used for the allylation of carbonyl compounds such as aldehydes, ketones, and imines. Homoallylic alcohols and amines are obtained via the C-C bond forming reaction. It can also be used to synthesize homoallylic +a-branched amines from aromatic and aliphatic aldehyde hydrazones, and ketone hydrazones.

Specifications

CAS 2551-83-9
Density 0.963 g/mL at 25°C
Boiling Point 146°C to 148°C
Molecular Formula C6H14O3Si
Linear Formula H2C=CHCH2Si(OCH3)3
Refractive Index n20/D 1.405
Quantity 5 mL
MDL Number MFCD00053865
Molecular Weight (g/mol) 162.26 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.