Learn More
Medchemexpress LLC Allyltrimethylsilane | 762-72-1 | MFCD00008635 | 99.6% | 114.26 g/mol | C6H14Si | 50 G
SDP
Catalog No. 5000301496
Encompass_Preferred
Allyltrimethylsilane is an organosilicon reagent (C6H14Si, MW 114.26 g/mol) commonly used as an allyl donor in organic synthesis, including Hosomi-Sakurai allylation and other carbon-carbon bond-forming reactions. It is supplied at research-grade purity with supporting documentation for laboratory handling and analysis.
  • High purity suitable for synthetic applications.
  • Effective allyl donor for Hosomi-Sakurai and related allylation reactions.
  • Liquid at ambient conditions for straightforward dosing.
  • Available in multiple pack sizes for small-scale or bulk needs.
  • Documentation provided: COA and SDS to support quality and safety.
Catalog No. 50-003-01496 Supplier Medchemexpress LLC Supplier No. HYW01982850G
Add to Cart
Add to Cart
Allyltrimethylsilane is an organosilicon reagent (C6H14Si, MW 114.26 g/mol) commonly used as an allyl donor in organic synthesis, including Hosomi-Sakurai allylation and other carbon-carbon bond-forming reactions. It is supplied at research-grade purity with supporting documentation for laboratory handling and analysis.
  • High purity suitable for synthetic applications.
  • Effective allyl donor for Hosomi-Sakurai and related allylation reactions.
  • Liquid at ambient conditions for straightforward dosing.
  • Available in multiple pack sizes for small-scale or bulk needs.
  • Documentation provided: COA and SDS to support quality and safety.