Learn More
Medchemexpress LLC Allyltrimethylsilane (trimethyl-prop-2-enylsilane) | 762-72-1 | MFCD00008635 | 99.6% | 114.26 g/mol | C6H14Si | 25 G
SDP
Catalog No. 5000304170
Encompass_Preferred
Allyltrimethylsilane (trimethyl-prop-2-enylsilane) is an organosilicon reagent used in organic synthesis to introduce allyl groups and as a silicon-containing building block. Supplied for research use with high purity, it is commonly employed in allylation and carbon-carbon bond-forming reactions under standard synthetic conditions.
  • High purity (≈99.6%) suitable for research applications
  • Useful reagent for allylation and C-C bond formation
  • Stable, easy-to-handle liquid for synthetic workflows
  • Available in multiple pack sizes to suit experimental scale
  • Intended for research use only
Catalog No. 50-003-04170 Supplier Medchemexpress LLC Supplier No. 25GHYW019828
Allyltrimethylsilane (trimethyl-prop-2-enylsilane) is an organosilicon reagent used in organic synthesis to introduce allyl groups and as a silicon-containing building block. Supplied for research use with high purity, it is commonly employed in allylation and carbon-carbon bond-forming reactions under standard synthetic conditions.
  • High purity (≈99.6%) suitable for research applications
  • Useful reagent for allylation and C-C bond formation
  • Stable, easy-to-handle liquid for synthetic workflows
  • Available in multiple pack sizes to suit experimental scale
  • Intended for research use only