Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allyltris(3,3,4,4,5,5,6,6,6-Nonafluorohexyl)Stannane

Catalog No. 439161GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-916-1GF 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 43-916-1GF Supplier Sigma Aldrich Supplier No. 439161GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Product of Fluorous Technologies, Inc. Protected by U.S. patents 6,156,896; 5,859,247; and 5,777,126

Specifications

CAS 14023-80-4
Molecular Formula C7H7IrO4
Linear Formula Ir(acac)(CO2)
MDL Number MFCD00049135
Quantity 250 mg
Synonym Allyltris(1H,1H,2H,2H-perfluorohexyl)stannane; Tris(1H,1H,2H,2H-perfluorohexyl)allyltin
Molecular Weight (g/mol) 347.35 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.