Learn More
Braintree Scientific Alm Retractor, 1/8 d, 2.75 spread, 2.25 prongs
SDP
Catalog No. 501954186
Encompass
Alm Retractor Specifications: Spread: 2.75" (7cm), Prongs: 2.75" (5.7cm), Depth: 0.125" (.32cm) **Sold for research purposes only and not for human use**
Catalog No. 50-195-4186 Supplier Braintree Scientific Supplier No. ALM112
Alm Retractor Specifications: Spread: 2.75" (7cm), Prongs: 2.75" (5.7cm), Depth: 0.125" (.32cm) **Sold for research purposes only and not for human use**