Learn More
Fine Science Tools ALM RETRACTOR W BLADES SS 7.5C
SDP
Catalog No. NC1270461
Encompass
Product: Alm Retractor with Blades/7.5cm/ Teeth: Atraumatic Blades/ Teeth Length: / Teeth Width: 5.5mm/ Teeth Depth: 2mm/ Maximum Spread: 4.5cm/ Alloy: Stainless Steel/ Length: 7.5cm/ Unit: Each
Catalog No. NC1270461 Supplier Fine Science Tools Supplier No. 1700908
Product: Alm Retractor with Blades/7.5cm/ Teeth: Atraumatic Blades/ Teeth Length: / Teeth Width: 5.5mm/ Teeth Depth: 2mm/ Maximum Spread: 4.5cm/ Alloy: Stainless Steel/ Length: 7.5cm/ Unit: Each