Learn More
World Precision Instrument Alm Self-Retaining Retractor, 7cm, 2x2 Prongs
SDP
Catalog No. 502521397
Encompass
  • 7 cm (2.75") Long
  • 2 x 2 Prongs
  • Maximum spread: 5.6 cm
  • Smaller, thinner prongs for use on mouse/rat eyelids
Catalog No. 50-252-1397 Supplier World Precision Instrument Supplier No. 501844
  • 7 cm (2.75") Long
  • 2 x 2 Prongs
  • Maximum spread: 5.6 cm
  • Smaller, thinner prongs for use on mouse/rat eyelids