Learn More
Medchemexpress LLC HY-17485 10mg Medchemexpress, Alminoprofen CAS:39718-89-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1748510MG
Encompass_Preferred
Medchemexpress, HY-17485 10mg Alminoprofen CAS:39718-89-3 Alminoprofen (EB-382) is a nonsteroidal anti-inflammatory drug (NSAID) of the phenylpropionic acid class. Alminoprofen possesses a dual anti-inflammatory action, by inhibiting both secretory phospholipase A2 (sPLA2) and COX-2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1965