Learn More
Medchemexpress LLC HY-107319 10mg Medchemexpress, Almitrine mesylate CAS:29608-49-9 Purity:>98%
SDP
Catalog No. 501968373
Encompass_Preferred
Medchemexpress, HY-107319 10mg Almitrine mesylate CAS:29608-49-9 Almitrine mesylate, a peripheral chemoreceptor agonist, inhibits selectively the Ca2+-dependent K+ channel. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8373 Supplier Medchemexpress LLC Supplier No. HY10731910MG
Medchemexpress, HY-107319 10mg Almitrine mesylate CAS:29608-49-9 Almitrine mesylate, a peripheral chemoreceptor agonist, inhibits selectively the Ca2+-dependent K+ channel. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.