Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Almitrine mesylate | 29608-49-9 | MFCD01679060 | 99.8% | 669.76 g·mol-1 | C28H37F2N7O6S2 | 100 MG
SDP

Catalog No. 5000240953
Encompass_Preferred

Almitrine mesylate is a research-grade peripheral chemoreceptor agonist that selectively inhibits Ca2+-dependent K+ channels, used in studies of ventilatory control and pulmonary vascular responses. Supplied as a high-purity salt with solubility and storage guidance for laboratory use.

  • High purity (99.8%).
  • Molecular weight 669.76 g·mol-1; formula C28H37F2N7O6S2.
  • Soluble in DMSO (≈100 mg/mL); in vivo vehicle protocols provided.
  • Suitable for in vitro and in vivo pharmacological studies.
  • Solid storage at 4°C; in solution store at -80°C up to 6 months.
  • Available in small research pack sizes and larger quantities by quote.

Catalog No. 50-002-40953 Supplier Medchemexpress LLC Supplier No. HY107319100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Almitrine mesylate is a research-grade peripheral chemoreceptor agonist that selectively inhibits Ca2+-dependent K+ channels, used in studies of ventilatory control and pulmonary vascular responses. Supplied as a high-purity salt with solubility and storage guidance for laboratory use.

  • High purity (99.8%).
  • Molecular weight 669.76 g·mol-1; formula C28H37F2N7O6S2.
  • Soluble in DMSO (≈100 mg/mL); in vivo vehicle protocols provided.
  • Suitable for in vitro and in vivo pharmacological studies.
  • Solid storage at 4°C; in solution store at -80°C up to 6 months.
  • Available in small research pack sizes and larger quantities by quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.