Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Almitrine mesylate | 29608-49-9 | MFCD01679060 | 99.8% | 669.8 g/mol | C28H37F2N7O6S2 | 1 ML
SDP

Catalog No. 5000240598
Encompass_Preferred

Almitrine mesylate is a research chemical that functions as a peripheral chemoreceptor agonist and selectively inhibits Ca2+-dependent K+ channels. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for laboratory research use only.

  • Acts as a peripheral chemoreceptor agonist.
  • Selectively inhibits Ca2+-dependent K+ channels.
  • Provided as a 10 mM solution in DMSO for experimental convenience.
  • High purity suitable for research assays.
  • Characterized by formula C28H37F2N7O6S2 and molecular weight 669.8 g/mol.

Catalog No. 50-002-40598 Supplier Medchemexpress LLC Supplier No. HY10731910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Almitrine mesylate is a research chemical that functions as a peripheral chemoreceptor agonist and selectively inhibits Ca2+-dependent K+ channels. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for laboratory research use only.

  • Acts as a peripheral chemoreceptor agonist.
  • Selectively inhibits Ca2+-dependent K+ channels.
  • Provided as a 10 mM solution in DMSO for experimental convenience.
  • High purity suitable for research assays.
  • Characterized by formula C28H37F2N7O6S2 and molecular weight 669.8 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.