Learn More
Selleck Chemical LLC Almitrine mesylate S4439-1g
SDP
Catalog No. 503110494
Encompass_Preferred
Almitrine mesylate (Almitrine bismesylate Almitrine dimethanesulfonate Almitrine dimesylate) a pharmacologically unique respiratory stimulant acts as an agonist of peripheral chemoreceptors located on the carotid bodies Almitrine mesylate inhibits the activity of Ca2 -dependent K channel by decreasing its open probability with IC50 of 0 22 M
Catalog No. 50-311-0494 Supplier Selleck Chemical LLC Supplier No. S44391G
Almitrine mesylate (Almitrine bismesylate Almitrine dimethanesulfonate Almitrine dimesylate) a pharmacologically unique respiratory stimulant acts as an agonist of peripheral chemoreceptors located on the carotid bodies Almitrine mesylate inhibits the activity of Ca2 -dependent K channel by decreasing its open probability with IC50 of 0 22 M