Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Almond Oil, Sweet, NF, Spectrum™ Chemical
SDP

Catalog No. 18600423 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-423 500 mL Amber Glass Bottle
18-600-422 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-600-423 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AL215500MLGL

missing translation for 'validMainframeMsg'

AL215, 8007-69-0

Spectrum™ Chemical Almond Oil, Sweet, NF is a natural, versatile and effective emollient known for its stability and high level of the "skin-nourishing" Linoleic Acid. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8007-69-0

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 mL
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.