Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Almonertinib (hydrochloride) | 2134096-03-8 | 98.9% | 562.11 | 50 MG
SDP

Catalog No. 5000438495
Encompass_Preferred

Almonertinib hydrochloride is an orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor. It exhibits high selectivity for EGFR-sensitizing and T790M resistance mutations and is utilized in non-small cell lung cancer research. It can also inhibit other EGFR sensitive mutations, including G719X, del19, L858R, and L861Q.

  • Orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor
  • High selectivity for EGFR-sensitizing and T790M resistance mutations
  • Potent inhibitory activity against T790M (IC50: 0.37 nM), T790M/L858R (IC50: 0.29 nM), and T790M/Del19 (IC50: 0.21 nM)
  • Less effective against wild-type EGFR (IC50: 3.39 nM)
  • Used for research in non-small cell lung cancer (NSCLC)
  • Inhibits other EGFR sensitive mutations such as G719X, del19, L858R, and L861Q

Catalog No. 50-004-38495 Supplier Medchemexpress LLC Supplier No. HY112823B50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Almonertinib hydrochloride is an orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor. It exhibits high selectivity for EGFR-sensitizing and T790M resistance mutations and is utilized in non-small cell lung cancer research. It can also inhibit other EGFR sensitive mutations, including G719X, del19, L858R, and L861Q.

  • Orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor
  • High selectivity for EGFR-sensitizing and T790M resistance mutations
  • Potent inhibitory activity against T790M (IC50: 0.37 nM), T790M/L858R (IC50: 0.29 nM), and T790M/Del19 (IC50: 0.21 nM)
  • Less effective against wild-type EGFR (IC50: 3.39 nM)
  • Used for research in non-small cell lung cancer (NSCLC)
  • Inhibits other EGFR sensitive mutations such as G719X, del19, L858R, and L861Q

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.