Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Almonertinib hydrochloride (HS-10296 hydrochloride) | 2134096-03-8 | 98.92% | 562.11 | 25 MG
SDP

Catalog No. 5000439418
Encompass_Preferred

Almonertinib hydrochloride is an orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor. It exhibits high selectivity for EGFR-sensitizing and T790M resistance mutations. It shows strong inhibitory activity against T790M, T790M/L858R, and T790M/Del19 (IC50: 0.37, 0.29, and 0.21 nM, respectively) and is less effective against the wild type (3.39 nM). It is used in the research of non-small cell lung cancer.

  • Orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor.
  • High selectivity for EGFR-sensitizing and T790M resistance mutations.
  • Strong inhibitory activity against T790M, T790M/L858R, and T790M/Del19.
  • Less effective against wild type.
  • Used in the research of non-small cell lung cancer.

Catalog No. 50-004-39418 Supplier Medchemexpress LLC Supplier No. 25MGHY112823B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Almonertinib hydrochloride is an orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor. It exhibits high selectivity for EGFR-sensitizing and T790M resistance mutations. It shows strong inhibitory activity against T790M, T790M/L858R, and T790M/Del19 (IC50: 0.37, 0.29, and 0.21 nM, respectively) and is less effective against the wild type (3.39 nM). It is used in the research of non-small cell lung cancer.

  • Orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor.
  • High selectivity for EGFR-sensitizing and T790M resistance mutations.
  • Strong inhibitory activity against T790M, T790M/L858R, and T790M/Del19.
  • Less effective against wild type.
  • Used in the research of non-small cell lung cancer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.