Learn More
Medchemexpress LLC Almonertinib mesylate | 2134096-06-1 | 99.67% | 621.75 | 100 MG
SDP
Catalog No. 5000437257
Encompass_Preferred
Almonertinib mesylate (HS-10296) is an orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor. It exhibits high selectivity for EGFR-sensitizing and T790M resistance mutations, showing significant inhibitory activity against T790M, T790M/L858R, and T790M/Del19 mutations while being less effective against wild-type EGFR. It is primarily used in research for non-small cell lung cancer and can also inhibit other EGFR sensitive mutations.
  • Orally available, irreversible EGFR tyrosine kinase inhibitor
  • High selectivity for EGFR-sensitizing and T790M resistance mutations
  • Significant inhibitory activity against various T790M mutations
  • Less effective against wild-type EGFR
  • Used in research for non-small cell lung cancer
  • Inhibits other EGFR sensitive mutations including G719X, del19, L858R, and L861Q
Catalog No. 50-004-37257 Supplier Medchemexpress LLC Supplier No. HY112823A100MG
Almonertinib mesylate (HS-10296) is an orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor. It exhibits high selectivity for EGFR-sensitizing and T790M resistance mutations, showing significant inhibitory activity against T790M, T790M/L858R, and T790M/Del19 mutations while being less effective against wild-type EGFR. It is primarily used in research for non-small cell lung cancer and can also inhibit other EGFR sensitive mutations.
  • Orally available, irreversible EGFR tyrosine kinase inhibitor
  • High selectivity for EGFR-sensitizing and T790M resistance mutations
  • Significant inhibitory activity against various T790M mutations
  • Less effective against wild-type EGFR
  • Used in research for non-small cell lung cancer
  • Inhibits other EGFR sensitive mutations including G719X, del19, L858R, and L861Q