Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Almonertinib (mesylate) | 2134096-06-1 | 99.7% | 621.75 | 1 ML
SDP

Catalog No. 5000438494
Encompass_Preferred

Almonertinib mesylate is an orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor. It shows high selectivity for EGFR-sensitizing and T790M resistance mutations, with inhibitory activity against T790M, T790M/L858R, and T790M/Del19. It is used for the research of non-small cell lung cancer (NSCLC). This compound can also inhibit other EGFR sensitive mutations, including G719X, del19, L858R, and L861Q.

  • Orally available inhibitor
  • Irreversible, third-generation EGFR tyrosine kinase inhibitor
  • High selectivity for EGFR-sensitizing and T790M resistance mutations
  • Inhibitory activity against T790M, T790M/L858R, and T790M/Del19
  • Less effective against wild type EGFR
  • Used for non-small cell lung cancer research
  • Can inhibit other EGFR sensitive mutations like G719X, del19, L858R, and L861Q

Catalog No. 50-004-38494 Supplier Medchemexpress LLC Supplier No. HY112823A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Almonertinib mesylate is an orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor. It shows high selectivity for EGFR-sensitizing and T790M resistance mutations, with inhibitory activity against T790M, T790M/L858R, and T790M/Del19. It is used for the research of non-small cell lung cancer (NSCLC). This compound can also inhibit other EGFR sensitive mutations, including G719X, del19, L858R, and L861Q.

  • Orally available inhibitor
  • Irreversible, third-generation EGFR tyrosine kinase inhibitor
  • High selectivity for EGFR-sensitizing and T790M resistance mutations
  • Inhibitory activity against T790M, T790M/L858R, and T790M/Del19
  • Less effective against wild type EGFR
  • Used for non-small cell lung cancer research
  • Can inhibit other EGFR sensitive mutations like G719X, del19, L858R, and L861Q

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.