Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Almonertinib mesylate | 2134096-06-1 | 99.7% | 621.75 | 25 MG
SDP

Catalog No. 5000439640
Encompass_Preferred

Almonertinib mesylate (HS-10296 mesylate) is an orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor with high selectivity for EGFR-sensitizing and T790M resistance mutations. It shows great inhibitory activity against T790M, T790M/L858R, and T790M/Del19 (IC50: 0.37, 0.29, and 0.21 nM, respectively), and is less effective against wild type (3.39 nM). It is used for the research of non-small cell lung cancer, and can also inhibit other EGFR sensitive mutations, including G719X, del19, L858R, and L861Q.

  • Orally available
  • Irreversible
  • Third-generation EGFR tyrosine kinase inhibitor
  • High selectivity for EGFR-sensitizing and T790M resistance mutations
  • Great inhibitory activity against T790M, T790M/L858R and T790M/Del19
  • Used for research of non-small cell lung cancer
  • Inhibits EGFR sensitive mutations (G719X, del19, L858R, L861Q)

Catalog No. 50-004-39640 Supplier Medchemexpress LLC Supplier No. 25MGHY112823A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Almonertinib mesylate (HS-10296 mesylate) is an orally available, irreversible, third-generation EGFR tyrosine kinase inhibitor with high selectivity for EGFR-sensitizing and T790M resistance mutations. It shows great inhibitory activity against T790M, T790M/L858R, and T790M/Del19 (IC50: 0.37, 0.29, and 0.21 nM, respectively), and is less effective against wild type (3.39 nM). It is used for the research of non-small cell lung cancer, and can also inhibit other EGFR sensitive mutations, including G719X, del19, L858R, and L861Q.

  • Orally available
  • Irreversible
  • Third-generation EGFR tyrosine kinase inhibitor
  • High selectivity for EGFR-sensitizing and T790M resistance mutations
  • Great inhibitory activity against T790M, T790M/L858R and T790M/Del19
  • Used for research of non-small cell lung cancer
  • Inhibits EGFR sensitive mutations (G719X, del19, L858R, L861Q)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.