Learn More
Medchemexpress LLC HY-10805 5mg Medchemexpress, Almorexant CAS:871224-64-5 Purity:>98%
SDP
Catalog No. 501968504
Encompass_Preferred
Medchemexpress, HY-10805 5mg Almorexant CAS:871224-64-5 Almorexant (ACT 078573) is a potent and competitive dual orexin 1 receptor (OX1)/orexin 2 receptor (OX2) antagonist with Ki values of 1.3 and 0.17 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8504 Supplier Medchemexpress LLC Supplier No. HY108055MG
Medchemexpress, HY-10805 5mg Almorexant CAS:871224-64-5 Almorexant (ACT 078573) is a potent and competitive dual orexin 1 receptor (OX1)/orexin 2 receptor (OX2) antagonist with Ki values of 1.3 and 0.17 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.