Learn More
Medchemexpress LLC HY-10805A 5mg Medchemexpress, Almorexant (hydrochloride) CAS:913358-93-7 Purity:>98%
SDP
Catalog No. 501968505
Encompass_Preferred
Medchemexpress, HY-10805A 5mg Almorexant (HCl) CAS:913358-93-7 Almorexant hydrochloride (ACT 078573 hydrochloride) is a potent and competitive dual orexin 1 receptor (OX1)/orexin 2 receptor (OX2) antagonist with Ki values of 1.3 and 0.17 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8505 Supplier Medchemexpress LLC Supplier No. HY10805A5MG
Medchemexpress, HY-10805A 5mg Almorexant (HCl) CAS:913358-93-7 Almorexant hydrochloride (ACT 078573 hydrochloride) is a potent and competitive dual orexin 1 receptor (OX1)/orexin 2 receptor (OX2) antagonist with Ki values of 1.3 and 0.17 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.