Learn More
Medchemexpress LLC Almorexant hydrochlo 100mg | 913358-93-7 | 100MG
SDP
Catalog No. 5000241728
Encompass_Preferred
Almorexant (ACT 078573) hydrochloride is an orally active potent and competitive dual orexin receptor antagonist with Kd values of 1 3 nM (OX1) and 0 17 nM (OX2) respectively Almorexant hydrochloride reversibly blocks signaling of orexin-A and orexin-B peptides Almorexant hydrochloride totally blocked the intracellular Ca2 signal pathway Almorexant hydrochloride stimulates caspase-3 activity in AsPC-1 cells and induces apoptosis[1 [2 [3 [4
Catalog No. 50-002-41728 Supplier Medchemexpress LLC Supplier No. HY10805A100MG
Almorexant (ACT 078573) hydrochloride is an orally active potent and competitive dual orexin receptor antagonist with Kd values of 1 3 nM (OX1) and 0 17 nM (OX2) respectively Almorexant hydrochloride reversibly blocks signaling of orexin-A and orexin-B peptides Almorexant hydrochloride totally blocked the intracellular Ca2 signal pathway Almorexant hydrochloride stimulates caspase-3 activity in AsPC-1 cells and induces apoptosis[1 [2 [3 [4