Learn More
Medchemexpress LLC Almorexant hydrochloride | 913358-93-7 | MFCD22419385 | 100.0% | 549.02 | C29H32ClF3N2O3 | 10 MG
SDP
Catalog No. 5000242968
Encompass_Preferred
Almorexant hydrochloride is an orally active, competitive dual orexin (OX1/OX2) receptor antagonist for research use. Provided as the hydrochloride salt, it is supplied in solid and solution formats with accompanying datasheet and safety documentation for laboratory studies of orexin signaling and sleep-wake regulation.
  • Orally active dual orexin receptor antagonist.
  • High reported purity for research applications.
  • Available as solid and as 10 mM solution in DMSO.
  • White to off-white solid appearance.
  • Includes datasheet and safety data sheet documentation.
  • Molecular weight 549.02 g/mol.
Catalog No. 50-002-42968 Supplier Medchemexpress LLC Supplier No. HY10805A10MG
Almorexant hydrochloride is an orally active, competitive dual orexin (OX1/OX2) receptor antagonist for research use. Provided as the hydrochloride salt, it is supplied in solid and solution formats with accompanying datasheet and safety documentation for laboratory studies of orexin signaling and sleep-wake regulation.
  • Orally active dual orexin receptor antagonist.
  • High reported purity for research applications.
  • Available as solid and as 10 mM solution in DMSO.
  • White to off-white solid appearance.
  • Includes datasheet and safety data sheet documentation.
  • Molecular weight 549.02 g/mol.