Learn More
Cayman Chemical Almorexnt-13C-d3 1mg
SDP
Supplier:  Cayman Chemical 290991 MG
Encompass_Preferred
An internal standard for the quantification of almorexant by GC- or LC-MS
Catalog No. 50-293-9465