Learn More
Cayman Chemical Almorexnt-13C-d3 500ug
SDP
Catalog No. 502940566
Encompass_Preferred
An internal standard for the quantification of almorexant by GC- or LC-MS
Catalog No. 50-294-0566 Supplier Cayman Chemical Supplier No. 29099500 UG
An internal standard for the quantification of almorexant by GC- or LC-MS