Learn More
Medchemexpress LLC Almotriptan hydrochloride-d6 | 1794782-57-2 | 1 MG
SDP
Catalog No. 5000407354
Encompass_Preferred
Almotriptan-d6 hydrochloride is a deuterium-labeled form of Almotriptan, which functions as a 5-HT1B/1D-receptor agonist primarily used in the treatment of migraine. The incorporation of stable heavy isotopes, such as deuterium, into drug molecules serves as a tracer for quantitation during the drug development process and can potentially influence the pharmacokinetic and metabolic profiles of the drugs.
  • Deuterium-labeled compound
  • Functions as a 5-HT1B/1D-receptor agonist
  • Used in the treatment of migraine
  • Serves as a tracer for quantitation in drug development
  • May affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-07354 Supplier Medchemexpress LLC Supplier No. HYB0383AS1MG
Almotriptan-d6 hydrochloride is a deuterium-labeled form of Almotriptan, which functions as a 5-HT1B/1D-receptor agonist primarily used in the treatment of migraine. The incorporation of stable heavy isotopes, such as deuterium, into drug molecules serves as a tracer for quantitation during the drug development process and can potentially influence the pharmacokinetic and metabolic profiles of the drugs.
  • Deuterium-labeled compound
  • Functions as a 5-HT1B/1D-receptor agonist
  • Used in the treatment of migraine
  • Serves as a tracer for quantitation in drug development
  • May affect pharmacokinetic and metabolic profiles