Learn More
Apexbio Technology LLC Almotriptan Malate 181183-52-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090822
Encompass_Preferred
Almotriptan Malate (CAS 181183-52-8) is a small molecule belonging to the triptan class acting as a selective agonist at 5-hydroxytryptamine 1B/1D (5-HT1B/1D) receptors By stimulating these receptors Almotriptan modulates cranial vasoconstriction and inhibits the release of pro-inflammatory neuropeptides biologically counteracting migraine pathophysiology In research applications Almotriptan Malate is utilized to investigate serotonergic signaling pathways migraine mechanisms and the pharmacodynamics of 5-HT1B/1D receptor modulation in neurological models
Catalog No. 50-000-90822 Supplier Apexbio Technology LLC Supplier No. B22435.1
Almotriptan Malate (CAS 181183-52-8) is a small molecule belonging to the triptan class acting as a selective agonist at 5-hydroxytryptamine 1B/1D (5-HT1B/1D) receptors By stimulating these receptors Almotriptan modulates cranial vasoconstriction and inhibits the release of pro-inflammatory neuropeptides biologically counteracting migraine pathophysiology In research applications Almotriptan Malate is utilized to investigate serotonergic signaling pathways migraine mechanisms and the pharmacodynamics of 5-HT1B/1D receptor modulation in neurological models