Learn More
Medchemexpress LLC Almotriptan (malate) | 181183-52-8 | 99.5% | 25 MG
SDP
Catalog No. 5000450989
Encompass_Preferred
Almotriptan malate is an orally active, highly selective 5-HT1B/1D-receptor agonist. It is utilized in research for treating migraine, inducing intracranial vasoconstriction, suppressing vasoactive peptides from trigeminal nerve endings, and inhibiting nociceptive neurotransmission in the trigeminocervical complex. This compound shows low nanomolar affinity for 5-HT1B and 5-HT1D receptors.
  • Highly selective 5-HT1B/1D-receptor agonist
  • Induces intracranial vasoconstriction
  • Suppresses vasoactive peptide release
  • Inhibits nociceptive neurotransmission
  • Exhibits low nanomolar affinity for 5-HT1B and 5-HT1D receptors
  • Has a mild antiemetic effect
  • Shows a slight, transient diuretic effect
Catalog No. 50-004-50989 Supplier Medchemexpress LLC Supplier No. 25MGHYB0383
Almotriptan malate is an orally active, highly selective 5-HT1B/1D-receptor agonist. It is utilized in research for treating migraine, inducing intracranial vasoconstriction, suppressing vasoactive peptides from trigeminal nerve endings, and inhibiting nociceptive neurotransmission in the trigeminocervical complex. This compound shows low nanomolar affinity for 5-HT1B and 5-HT1D receptors.
  • Highly selective 5-HT1B/1D-receptor agonist
  • Induces intracranial vasoconstriction
  • Suppresses vasoactive peptide release
  • Inhibits nociceptive neurotransmission
  • Exhibits low nanomolar affinity for 5-HT1B and 5-HT1D receptors
  • Has a mild antiemetic effect
  • Shows a slight, transient diuretic effect