Learn More
Selleck Chemical LLC Almotriptan Malate S2096-10mg
SDP
Catalog No. 503111127
Encompass_Preferred
Almotriptan Malate (LAS 31416) is a selective 5-hydroxytryptamine1B/1D (5-HT1B/1D) receptor agonist used for the treatment of Migraine attacks in adults
Catalog No. 50-311-1127 Supplier Selleck Chemical LLC Supplier No. S209610MG
Almotriptan Malate (LAS 31416) is a selective 5-hydroxytryptamine1B/1D (5-HT1B/1D) receptor agonist used for the treatment of Migraine attacks in adults