Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Almotriptan malate

Catalog No. AC465402500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465402500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC465402500 Supplier Thermo Scientific Chemicals Supplier No. 465402500
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181183-52-8
Molecular Formula C21H31N3O7S
Molecular Weight (g/mol) 469.55
InChI Key QHATUKWEVNMHRY-UHFFFAOYNA-N
IUPAC Name 2-hydroxybutanedioic acid; dimethyl(2-{5-[(pyrrolidine-1-sulfonyl)methyl]-1H-indol-3-yl}ethyl)amine
SMILES OC(CC(O)=O)C(O)=O.CN(C)CCC1=CNC2=CC=C(CS(=O)(=O)N3CCCC3)C=C12

Specifications

Quantity 250 mg
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 469.55
Chemical Name or Material Almotriptan malate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.