Learn More
Supertek™ Scientific Alnico Bar Magnet Set
GREENER_CHOICESCI_ED
Catalog No. S82805 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
3+
Catalog No. Grade
S82805 3+
1 options
This item is not returnable. View return policy
Catalog No. S82805 Supplier Supertek™ Scientific Supplier No. PH40023A/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Scientific Alnico Bar Magnet Set has unlimited uses
  • Magnet bars painted red with pole indicators in white
  • Magnet Set
Specifications
Material Alnico
Shape Rectangular
Grade 3+
Product Type Magnet
Includes Separator and keepers
Dimensions (L x W x H) 2 x 0.5 x 0.25 in.