Learn More
Cayman Chemical Alantolactone, Purity: ≥98%, CAS: 546-43-0, 10 mg
SDP
Catalog No. 502988560
Encompass_Preferred
A sesquiterpene lactone with diverse biological activities; inhibits LPS-induced increases in nitric oxide, IL-6, and TNF-α production in RAW 246.7 cells by 76.09, 81.54, and 71.23%, respectively, at 10 μM; decreases cell viability, GSH levels, and the mitochondrial membrane potential, induces apoptosis, and increases ROS generation in HepG2 cells at 40 μM; inhibits phosphorylation and nuclear translocation of STAT3 in MDA-MB-231 cells; reduces tumor growth in an MDA-MB-231 mouse xenograft model at 2.5 mg/kg
Catalog No. 50-298-8560 Supplier Cayman Chemical Supplier No. 2976210 MG
A sesquiterpene lactone with diverse biological activities; inhibits LPS-induced increases in nitric oxide, IL-6, and TNF-α production in RAW 246.7 cells by 76.09, 81.54, and 71.23%, respectively, at 10 μM; decreases cell viability, GSH levels, and the mitochondrial membrane potential, induces apoptosis, and increases ROS generation in HepG2 cells at 40 μM; inhibits phosphorylation and nuclear translocation of STAT3 in MDA-MB-231 cells; reduces tumor growth in an MDA-MB-231 mouse xenograft model at 2.5 mg/kg